Assay Details
| Gene Symbol: C20orf173 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015298107.2 | XP_015153593.1 | 9 - 10 | 728 | 75 | |
| XM_015298108.2 | XP_015153594.1 | 9 - 10 | 944 | 75 | ||
| XM_025143326.1 | XP_024999094.1 | 7 - 8 | 607 | 75 | ||
| XM_417860.5 | XP_417860.3 | 9 - 10 | 735 | 75 | ||
| GenBank mRNA | AJ866777.1 | - | 6 - 7 | 429 | 75 | |
More Information
Set Membership: |
|

C20orf173
Genomic Map