Assay Details
| Gene Symbol: C20orf173 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015298107.2 | XP_015153593.1 | 10 - 11 | 911 | 96 | |
| XM_015298108.2 | XP_015153594.1 | 10 - 11 | 1127 | 96 | ||
| XM_025143326.1 | XP_024999094.1 | 8 - 9 | 790 | 96 | ||
| XM_417860.5 | XP_417860.3 | 10 - 11 | 918 | 96 | ||
| GenBank mRNA | AJ866777.1 | - | 7 - 8 | 612 | 96 | |
| JX035870.1 | - | 7 - 8 | 641 | 96 | ||
More Information
Set Membership: |
|

C20orf173
Genomic Map