Assay Details
| Gene Symbol: CKAP2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015297503.2 | XP_015152989.1 | 5 - 6 | 1573 | 87 | |
| XM_424317.6 | XP_424317.4 | 6 - 7 | 1644 | 87 | ||
| GenBank mRNA | CR386920.1 | - | 1 - 2 | 77 | 87 | |
More Information
Set Membership: |
|

CKAP2L
Genomic Map