Assay Details
| Gene Symbol: LARP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015278970.2 | XP_015134456.1 | 2 - 3 | 811 | 77 | |
| XM_015278971.2 | XP_015134457.1 | 3 - 4 | 1549 | 77 | ||
| XM_015278972.2 | XP_015134458.1 | 3 - 4 | 1314 | 77 | ||
| XM_015278973.2 | XP_015134459.1 | 2 - 3 | 1814 | 77 | ||
| XM_015278974.2 | XP_015134460.1 | 2 - 3 | 1212 | 77 | ||
| GenBank mRNA | CR390427.1 | - | 1 - 2 | 80 | 77 | |
More Information
Set Membership: |
|

LARP6
Genomic Map