Assay Details
| Gene Symbol: MED21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_416442.6 | XP_416442.2 | 2 - 3 | 195 | 73 | |
| GenBank mRNA | CR390891.1 | - | 2 - 3 | 171 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MED21| Gene Symbol: MED21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_416442.6 | XP_416442.2 | 2 - 3 | 195 | 73 | |
| GenBank mRNA | CR390891.1 | - | 2 - 3 | 171 | 73 | |
Set Membership: |
|