Assay Details
| Gene Symbol: ITGBL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_416966.5 | XP_416966.3 | 1 - 2 | 423 | 103 | |
| GenBank mRNA | CR405873.1 | - | 1 - 2 | 71 | 103 | |
More Information
Set Membership: |
|

ITGBL1
Genomic Map