Assay Details
| Gene Symbol: SHF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015279099.2 | XP_015134585.1 | 6 - 7 | 1436 | 75 | |
| XM_015279101.2 | XP_015134587.1 | 5 - 6 | 1298 | 75 | ||
| GenBank mRNA | CR407298.1 | - | 2 - 3 | 261 | 75 | |
More Information
Set Membership: |
|

SHF
Genomic Map