Assay Details
| Gene Symbol: MAN1A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_419762.6 | XP_419762.5 | 5 - 6 | 1130 | 63 | |
| GenBank mRNA | CR407310.1 | - | 5 - 6 | 438 | 63 | |
More Information
Set Membership: |
|

MAN1A1
Genomic Map