Assay Details
| Gene Symbol: OVOL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_415018.6 | XP_415018.4 | 2 - 3 | 843 | 87 | |
| GenBank mRNA | CR522981.1 | - | 1 - 2 | 200 | 87 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

OVOL3| Gene Symbol: OVOL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_415018.6 | XP_415018.4 | 2 - 3 | 843 | 87 | |
| GenBank mRNA | CR522981.1 | - | 1 - 2 | 200 | 87 | |
Set Membership: |
|