Assay Details
| Gene Symbol: AHCY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_417331.6 | XP_417331.5 | 10 | 110 | ||
| GenBank mRNA | CR523289.1 | - | 1 | 846 | 110 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AHCY| Gene Symbol: AHCY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_417331.6 | XP_417331.5 | 10 | 110 | ||
| GenBank mRNA | CR523289.1 | - | 1 | 846 | 110 | |
Set Membership: |
|