Assay Details
| Gene Symbol: C9ORF58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_415461.6 | XP_415461.1 | 3 - 4 | 215 | 94 | |
| GenBank mRNA | CR523365.1 | - | 3 - 4 | 174 | 94 | |
| KJ101611.1 | - | 3 - 4 | 180 | 94 | ||
More Information
Set Membership: |
|

C9ORF58
Genomic Map