Assay Details
| Gene Symbol: RHOGL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | CR733247.1 | - | 7 - 8 | 722 | 81 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RHOGL| Gene Symbol: RHOGL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | CR733247.1 | - | 7 - 8 | 722 | 81 | |
Set Membership: |
|