Assay Details
Gene Symbol: CYP26C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015288845.2 | XP_015144331.1 | 4 | 800 | 59 | |
XM_421678.6 | XP_421678.5 | 5 | 1076 | 59 | ||
GenBank mRNA | CR406968.1 | - | 1 | 48 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
