Assay Details
Gene Symbol: LOC101747443 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025145870.1 | XP_025001638.1 | 3 | 349 | 109 | |
GenBank mRNA | CR523517.1 | - | 4 | 850 | 109 |
Gene Symbol: LOC112531066 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025145977.1 | XP_025001745.1 | 3 | 224 | 109 |
Gene Symbol: ZNF555 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015273175.2 | XP_015128661.2 | 5 | 1439 | 109 | |
GenBank mRNA | BX934716.2 | - | 2 | 209 | 109 | |
BX935080.1 | - | 5 | 488 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
