Assay Details
| Gene Symbol: RBM7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006318.1 | NP_001006318.1 | 1 - 2 | 139 | 112 | |
| XM_015297995.2 | XP_015153481.1 | 1 - 2 | 169 | 112 | ||
| GenBank mRNA | AJ719791.1 | - | 1 - 2 | 139 | 112 | |
More Information
Set Membership: |
|

RBM7
Genomic Map