Assay Details
| Gene Symbol: SDHD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006321.1 | NP_001006321.1 | 3 - 4 | 336 | 73 | |
| GenBank mRNA | AJ720714.1 | - | 3 - 4 | 336 | 73 | |
| CR405813.1 | - | 3 - 4 | 316 | 73 | ||
More Information
Set Membership: |
|

SDHD
Genomic Map