Assay Details
Gene Symbol: SIKE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006324.2 | NP_001006324.2 | 709 | 124 | ||
GenBank mRNA | AJ720809.1 | - | 711 | 124 | ||
BU133794.1 | - | 322 | 124 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
