Assay Details
| Gene Symbol: NFYA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006325.1 | NP_001006325.1 | 725 | 76 | ||
| XM_015298937.2 | XP_015154423.1 | 1086 | 76 | |||
| XM_015298938.2 | XP_015154424.1 | 2624 | 76 | |||
| XM_015298939.2 | XP_015154425.1 | 1480 | 76 | |||
| XM_015298940.2 | XP_015154426.1 | 1083 | 76 | |||
| XM_015298941.2 | XP_015154427.1 | 1083 | 76 | |||
| XM_015298942.2 | XP_015154428.1 | 1080 | 76 | |||
| XM_015298943.2 | XP_015154429.1 | 1068 | 76 | |||
| XM_015298944.2 | XP_015154430.1 | 864 | 76 | |||
| XM_015298945.2 | XP_015154431.1 | 981 | 76 | |||
| XM_025143700.1 | XP_024999468.1 | 1104 | 76 | |||
| XM_025143701.1 | XP_024999469.1 | 1101 | 76 | |||
| XM_025143702.1 | XP_024999470.1 | 1101 | 76 | |||
| XM_025143703.1 | XP_024999471.1 | 1098 | 76 | |||
| XM_025143704.1 | XP_024999472.1 | 2622 | 76 | |||
| XM_025143705.1 | XP_024999473.1 | 2535 | 76 | |||
| XM_025143706.1 | XP_024999474.1 | 890 | 76 | |||
| GenBank mRNA | AJ719407.1 | - | 725 | 76 | ||
More Information
Set Membership: |
|

NFYA
Genomic Map