Assay Details
| Gene Symbol: RPL7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006345.2 | NP_001006345.1 | 3 - 4 | 314 | 70 | |
| GenBank mRNA | AJ720578.1 | - | 3 - 4 | 308 | 70 | |
| BU135171.1 | - | 3 - 4 | 314 | 70 | ||
| CR338953.1 | - | 3 - 4 | 294 | 70 | ||
| CR352384.1 | - | 2 - 3 | 284 | 70 | ||
| CR353058.1 | - | 3 - 4 | 291 | 70 | ||
More Information
Set Membership: |
|

RPL7
Genomic Map