Assay Details
| Gene Symbol: FABP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006346.1 | NP_001006346.1 | 2 - 3 | 300 | 111 | |
| GenBank mRNA | AJ720717.1 | - | 2 - 3 | 300 | 111 | |
| BX936126.1 | - | 2 - 3 | 469 | 111 | ||
More Information
Set Membership: |
|

FABP5
Genomic Map