Assay Details
| Gene Symbol: FABP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006346.1 | NP_001006346.1 | 3 - 4 | 401 | 112 | |
| GenBank mRNA | AJ720717.1 | - | 3 - 4 | 401 | 112 | |
| BX933190.2 | - | 1 - 2 | 698 | 112 | ||
| BX936126.1 | - | 3 - 4 | 570 | 112 | ||
More Information
Set Membership: |
|

FABP5
Genomic Map