Assay Details
| Gene Symbol: OTUD6A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006347.1 | NP_001006347.1 | 4 - 5 | 696 | 93 | |
| GenBank mRNA | AJ720738.1 | - | 4 - 5 | 696 | 93 | |
More Information
Set Membership: |
|

OTUD6A
Genomic Map