Assay Details
| Gene Symbol: ARF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006352.1 | NP_001006352.1 | 4 - 5 | 518 | 90 | |
| GenBank mRNA | AJ719484.1 | - | 4 - 5 | 518 | 90 | |
More Information
Set Membership: |
|

ARF1
Genomic Map