Assay Details
| Gene Symbol: FH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006382.1 | NP_001006382.1 | 2 - 3 | 310 | 80 | |
| GenBank mRNA | AJ719803.1 | - | 2 - 3 | 310 | 80 | |
More Information
Set Membership: |
|

FH
Genomic Map