Assay Details
| Gene Symbol: TGS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006389.1 | NP_001006389.1 | 5 - 6 | 1355 | 74 | |
| GenBank mRNA | AJ720557.1 | - | 5 - 6 | 1355 | 74 | |
More Information
Set Membership: |
|

TGS1
Genomic Map