Assay Details
| Gene Symbol: LYN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006390.1 | NP_001006390.1 | 2 - 3 | 186 | 63 | |
| GenBank mRNA | AJ719465.1 | - | 2 - 3 | 186 | 63 | |
| AJ720258.1 | - | 2 - 3 | 239 | 63 | ||
More Information
Set Membership: |
|

LYN
Genomic Map