Assay Details
| Gene Symbol: FAM49A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006418.2 | NP_001006418.1 | 6 - 7 | 537 | 75 | |
| XM_015276050.2 | XP_015131536.1 | 4 - 5 | 432 | 75 | ||
| XM_015276051.2 | XP_015131537.1 | 6 - 7 | 591 | 75 | ||
| XM_015276053.2 | XP_015131539.1 | 6 - 7 | 610 | 75 | ||
| XM_015276054.2 | XP_015131540.1 | 7 - 8 | 684 | 75 | ||
| XM_015276055.2 | XP_015131541.1 | 8 - 9 | 763 | 75 | ||
| XM_015276056.2 | XP_015131542.1 | 6 - 7 | 601 | 75 | ||
| XM_015276060.2 | XP_015131546.1 | 5 - 6 | 507 | 75 | ||
| XM_025148784.1 | XP_025004552.1 | 7 - 8 | 690 | 75 | ||
| XM_025148785.1 | XP_025004553.1 | 6 - 7 | 610 | 75 | ||
| XM_025148786.1 | XP_025004554.1 | 5 - 6 | 530 | 75 | ||
| GenBank mRNA | AJ720980.1 | - | 6 - 7 | 538 | 75 | |
More Information
Set Membership: |
|

FAM49A
Genomic Map