Assay Details
| Gene Symbol: MCM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006421.1 | NP_001006421.1 | 6 - 7 | 1007 | 62 | |
| GenBank mRNA | AJ719352.1 | - | 6 - 7 | 1007 | 62 | |
More Information
Set Membership: |
|

MCM3
Genomic Map