Assay Details
| Gene Symbol: CHMP1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006428.1 | NP_001006428.1 | 3 - 4 | 325 | 66 | |
| GenBank mRNA | AJ719963.1 | - | 3 - 4 | 325 | 66 | |
| CR387811.1 | - | 3 - 4 | 342 | 66 | ||
More Information
Set Membership: |
|

CHMP1B
Genomic Map