Assay Details
| Gene Symbol: CHMP1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006428.1 | NP_001006428.1 | 7 - 8 | 619 | 77 | |
| GenBank mRNA | AJ719963.1 | - | 7 - 8 | 619 | 77 | |
| CR387811.1 | - | 7 - 8 | 636 | 77 | ||
More Information
Set Membership: |
|

CHMP1B
Genomic Map