Assay Details
| Gene Symbol: APOOL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006434.2 | NP_001006434.1 | 6 - 7 | 530 | 71 | |
| XM_025149563.1 | XP_025005331.1 | 6 - 7 | 515 | 71 | ||
| GenBank mRNA | AJ720229.1 | - | 6 - 7 | 531 | 71 | |
More Information
Set Membership: |
|

APOOL
Genomic Map