Assay Details
| Gene Symbol: AGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006445.1 | NP_001006445.1 | 2 - 3 | 279 | 63 | |
| GenBank mRNA | AJ720474.1 | - | 2 - 3 | 279 | 63 | |
More Information
Set Membership: |
|

AGA
Genomic Map