Assay Details
| Gene Symbol: CNOT7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006454.1 | NP_001006454.1 | 6 - 7 | 849 | 95 | |
| XM_015285436.2 | XP_015140922.1 | 6 - 7 | 966 | 95 | ||
| XM_025149692.1 | XP_025005460.1 | 6 - 7 | 1185 | 95 | ||
| GenBank mRNA | AJ720325.1 | - | 6 - 7 | 849 | 95 | |
More Information
Set Membership: |
|

CNOT7
Genomic Map