Assay Details
| Gene Symbol: MXD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006460.1 | NP_001006460.1 | 2 - 3 | 306 | 69 | |
| XM_015285898.2 | XP_015141384.1 | 2 - 3 | 370 | 69 | ||
| XM_015285899.2 | XP_015141385.1 | 2 - 3 | 317 | 69 | ||
| GenBank mRNA | AJ719815.1 | - | 2 - 3 | 306 | 69 | |
More Information
Set Membership: |
|

MXD4
Genomic Map