Assay Details
| Gene Symbol: RRAS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006466.1 | NP_001006466.1 | 2 - 3 | 418 | 86 | |
| XM_015286388.2 | XP_015141874.1 | 2 - 3 | 134 | 86 | ||
| XM_015286389.1 | XP_015141875.1 | 2 - 3 | 128 | 86 | ||
| XM_015286390.2 | XP_015141876.1 | 2 - 3 | 109 | 86 | ||
| GenBank mRNA | AJ720666.1 | - | 2 - 3 | 418 | 86 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015286390.2 transcripts.
Set Membership: |
|

RRAS
Genomic Map