Assay Details
| Gene Symbol: SRSF5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006476.2 | NP_001006476.2 | 3 - 4 | 356 | 104 | |
| XM_015287323.2 | XP_015142809.1 | 4 - 5 | 4316 | 104 | ||
| XM_025150719.1 | XP_025006487.1 | 4 - 5 | 4581 | 104 | ||
| GenBank mRNA | AJ720000.1 | - | 3 - 4 | 357 | 104 | |
More Information
Set Membership: |
|

SRSF5A
Genomic Map