Assay Details
| Gene Symbol: SRSF5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006476.2 | NP_001006476.2 | 5 - 6 | 511 | 86 | |
| XM_015287323.2 | XP_015142809.1 | 6 - 7 | 4471 | 86 | ||
| XM_025150719.1 | XP_025006487.1 | 6 - 7 | 4736 | 86 | ||
| GenBank mRNA | AJ720000.1 | - | 5 - 6 | 512 | 86 | |
More Information
Set Membership: |
|

SRSF5A
Genomic Map