Assay Details
| Gene Symbol: SRP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006478.2 | NP_001006478.1 | 4 - 5 | 296 | 72 | |
| GenBank mRNA | AJ720841.1 | - | 4 - 5 | 297 | 72 | |
More Information
Set Membership: |
|

SRP14
Genomic Map