Assay Details
| Gene Symbol: ZNF410 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006480.1 | NP_001006480.1 | 6 - 7 | 939 | 64 | |
| XM_015287385.2 | XP_015142871.1 | 6 - 7 | 963 | 64 | ||
| GenBank mRNA | AJ720956.1 | - | 6 - 7 | 939 | 64 | |
More Information
Set Membership: |
|

ZNF410
Genomic Map