Assay Details
| Gene Symbol: HOPX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204556.1 | NP_989887.1 | 2 - 3 | 260 | 54 | |
| XM_015276108.2 | XP_015131594.1 | 3 - 4 | 3797 | 54 | ||
| XM_015276109.2 | XP_015131595.1 | 1 - 2 | 188 | 54 | ||
| XM_025149488.1 | XP_025005256.1 | 4 - 5 | 4112 | 54 | ||
| XM_025149489.1 | XP_025005257.1 | 3 - 4 | 4030 | 54 | ||
| XM_025149490.1 | XP_025005258.1 | 2 - 3 | 3715 | 54 | ||
| XM_025149491.1 | XP_025005259.1 | 2 - 3 | 189 | 54 | ||
| GenBank mRNA | AF492684.1 | - | 2 - 3 | 260 | 54 | |
| BX931425.1 | - | 3 - 4 | 754 | 54 | ||
| BX934519.1 | - | 2 - 3 | 250 | 54 | ||
| BX935796.1 | - | 1 - 2 | 418 | 54 | ||
| BX935939.1 | - | 235 | 54 | |||
| CR406220.1 | - | 1 - 2 | 245 | 54 | ||
More Information
Set Membership: |
|

HOPX
Genomic Map