Assay Details
| Gene Symbol: FKBP1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204567.1 | NP_989898.1 | 3 - 4 | 249 | 80 | |
| XM_015284949.2 | XP_015140435.1 | 3 - 4 | 277 | 80 | ||
| GenBank mRNA | AB074887.1 | - | 3 - 4 | 249 | 80 | |
More Information
Set Membership: |
|

FKBP1B
Genomic Map