Assay Details
| Gene Symbol: CPZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204578.4 | NP_989909.3 | 10 - 11 | 1674 | 63 | |
| GenBank mRNA | AF351205.1 | - | 10 - 11 | 1592 | 63 | |
More Information
Set Membership: |
|

CPZ
Genomic Map