Assay Details
| Gene Symbol: SSB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193669.1 | NP_001180598.1 | 2 - 3 | 348 | 97 | |
| XM_015289615.2 | XP_015145101.1 | 2 - 3 | 325 | 97 | ||
| XM_025152085.1 | XP_025007853.1 | 2 - 3 | 295 | 97 | ||
| GenBank mRNA | AF467897.1 | - | 1 - 2 | 175 | 97 | |
| AJ449190.1 | - | 2 - 3 | 349 | 97 | ||
| AJ720013.1 | - | 2 - 3 | 325 | 97 | ||
| BU379542.1 | - | 2 - 3 | 299 | 97 | ||
More Information
Set Membership: |
|

SSB
Genomic Map