Assay Details
| Gene Symbol: PARK7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204585.1 | NP_989916.1 | 5 - 6 | 460 | 66 | |
| XM_015296819.2 | XP_015152305.1 | 5 - 6 | 399 | 66 | ||
| GenBank mRNA | AB076264.1 | - | 5 - 6 | 460 | 66 | |
| HM012714.1 | - | 4 - 5 | 339 | 66 | ||
More Information
Set Membership: |
|

PARK7
Genomic Map