Assay Details
| Gene Symbol: BIRC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204588.2 | NP_989919.1 | 5 - 6 | 1161 | 81 | |
| XM_015278182.2 | XP_015133668.1 | 5 - 6 | 1192 | 81 | ||
| XM_015278183.2 | XP_015133669.1 | 5 - 6 | 1180 | 81 | ||
| GenBank mRNA | AF451854.1 | - | 5 - 6 | 1162 | 81 | |
More Information
Set Membership: |
|

BIRC8
Genomic Map