Assay Details
| Gene Symbol: NR2E3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204594.1 | NP_989925.1 | 2 - 3 | 221 | 98 | |
| XM_025153796.1 | XP_025009564.1 | 1 - 2 | 251 | 98 | ||
| GenBank mRNA | AY053465.1 | - | 2 - 3 | 221 | 98 | |
More Information
Set Membership: |
|

NR2E3
Genomic Map