Assay Details
| Gene Symbol: EIF2S2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204597.1 | NP_989928.1 | 6 - 7 | 764 | 110 | |
| GenBank mRNA | AJ294707.1 | - | 6 - 7 | 764 | 110 | |
| CR385806.1 | - | 6 - 7 | 773 | 110 | ||
More Information
Set Membership: |
|

EIF2S2
Genomic Map