Assay Details
| Gene Symbol: SP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204603.1 | NP_989934.1 | 6 - 7 | 2203 | 74 | |
| XM_015289377.2 | XP_015144863.1 | 7 - 8 | 2316 | 74 | ||
| XM_025152088.1 | XP_025007856.1 | 4 - 5 | 2021 | 74 | ||
| GenBank mRNA | AJ317961.1 | - | 6 - 7 | 2203 | 74 | |
| AJ719471.1 | - | 4 - 5 | 1851 | 74 | ||
More Information
Set Membership: |
|

SP3
Genomic Map