Assay Details
| Gene Symbol: NFIL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204618.2 | NP_989949.1 | 2 | 1541 | 65 | |
| XM_015280234.2 | XP_015135720.1 | 2 | 1582 | 65 | ||
| XM_015280235.1 | XP_015135721.1 | 2 | 1428 | 65 | ||
| GenBank mRNA | AF335427.1 | - | 1 | 1251 | 65 | |
| DR432119.1 | - | 1 | 229 | 65 | ||
More Information
Set Membership: |
|

NFIL3
Genomic Map