Assay Details
| Gene Symbol: MOXD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204624.1 | NP_989955.1 | 4 - 5 | 840 | 62 | |
| GenBank mRNA | AF327450.1 | - | 4 - 5 | 840 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MOXD1| Gene Symbol: MOXD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204624.1 | NP_989955.1 | 4 - 5 | 840 | 62 | |
| GenBank mRNA | AF327450.1 | - | 4 - 5 | 840 | 62 | |
Set Membership: |
|